EMAIL THIS PAGE TO A FRIEND
Journal of cutaneous pathology
Cutaneous clear cell squamous cell carcinoma in situ : clinical, histological and immunohistochemical characterization.
PMID 17302606
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB5488 Anti-CRABP2 Antibody, ascites fluid, clone, Chemicon®