EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Enantioseparation of 1-phenyl-1-propanol on cellulose-derived chiral stationary phase by supercritical fluid chromatography II. Non-linear isotherm.
PMID 17303143
Abstract
The separation of the enantiomers of 1-phenyl-1-propanol by supercritical fluid chromatography on a chiral stationary phase, which consists of cellulose tris (3,5-dimethylphenylcarbamate) coated on a silica support (Chiralcel-OD), is studied under overloaded, non-linear chromatographic conditions. Pulse experiments are performed at a temperature of 30 degrees C using supercritical CO(2) modified with methanol as a mobile phase. The parameters of the binary Langmuir adsorption isotherm are determined by the inverse method, comparing experimental and simulated peak responses. Isotherm parameters are derived for modifier concentrations between 1 and 5% (w/w) and operating pressures between 125 and 185 bar, and the dependency of the isotherm parameters, namely the Henry constant and the saturation capacity, on operating conditions is investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
111309
1-Phenyl-1-propanol, ≥97%
C9H12O
W288403
1-Phenyl-1-propanol, ≥97%, FG
C9H12O