EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Two-photon uncaging with the efficient 3,5-dibromo-2,4-dihydroxycinnamic caging group.
PMID 17310488
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
168637
2,4-Dihydroxybenzaldehyde, 98%
C7H6O3