EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Ligand accelerated indium(III)-catalyzed asymmetric alkynylation of aldehydes with 2-methyl-3-butyn-2-ol as an ethyne equivalent donor.
PMID 17311130
Abstract
Indium(III)-catalyzed asymmetric alkynylation of aryl, heteroaryl, alkyl and alkenyl aldehydes with 2-methyl-3-butyn-2-ol as an ethyne equivalent donor was realized, and products were obtained in moderate to good yields (up to 97%) and high enantioselectivities (up to 99% ee) using 2-10 mol% of catalyst.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129763
2-Methyl-3-butyn-2-ol, 98%
C5H8O