EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
Influence of translation efficiency of homologous viral proteins on the endogenous presentation of CD8+ T cell epitopes.
PMID 17312009
Abstract
A significant proportion of endogenously processed CD8(+) T cell epitopes are derived from newly synthesized proteins and rapidly degrading polypeptides (RDPs). It has been hypothesized that the generation of rapidly degrading polypeptides and CD8(+) T cell epitopes from these RDP precursors may be influenced by the efficiency of protein translation. Here we address this hypothesis by using the Epstein-Barr virus-encoded nuclear antigen 1 protein (EBNA1), with or without its internal glycine-alanine repeat sequence (EBNA1 and EBNA1DeltaGA, respectively), which display distinct differences in translation efficiency. We demonstrate that RDPs constitute a significant proportion of newly synthesized EBNA1 and EBNA1DeltaGA and that the levels of RDPs produced by each of these proteins directly correlate with the translation efficiency of either EBNA1 or EBNA1DeltaGA. As a consequence, a higher number of major histocompatibility complex-peptide complexes can be detected on the surface of cells expressing EBNA1DeltaGA, and these cells are more efficiently recognized by virus-specific cytotoxic T lymphocytes compared to the full-length EBNA1. More importantly, we also demonstrate that the endogenous processing of these CD8(+) T cell epitopes is predominantly determined by the rate at which the RDPs are generated rather than the intracellular turnover of these proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50150
Gly-Ala, ≥99.0% (NT)
C5H10N2O3