EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Apoptotic natural cell death in developing primate dopamine midbrain neurons occurs during a restricted period in the second trimester of gestation.
PMID 17313945