EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design, synthesis, and activity of 2-imidazol-1-ylpyrimidine derived inducible nitric oxide synthase dimerization inhibitors.
PMID 17315988
Abstract
By the screening of a combinatorial library for inhibitors of nitric oxide (NO) formation by the inducible isoform of nitric oxide synthase (iNOS) using a whole-cell assay, 2-(imidazol-1-yl)pyrimidines were identified. Compounds were found to inhibit the dimerization of iNOS monomers, thus preventing the formation of the dimeric, active form of the enzyme. Optimization led to the selection of the potent, selective, and orally available iNOS dimerization inhibitor, 21b, which significantly ameliorated adjuvant-induced arthritis in a rat model. Analysis of the crystal structure of the 21b--iNOS monomer complex provided a rationalization for both the SAR and the mechanism by which 21b blocks the formation of the protein--protein interaction present in the dimeric form of iNOS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
145289
4-Chloro-2-methylthiopyrimidine, 98%
C5H5ClN2S