EMAIL THIS PAGE TO A FRIEND
Nephrology (Carlton, Vic.)
The CARI guidelines. Kidney stones: cystine stones.
PMID 17316277
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1323
L-Cystine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12N2O4S2