EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
A matrix metalloproteinase-9 activation cascade by hepatic stellate cells in trans-differentiation in the three-dimensional extracellular matrix.
PMID 17322299