EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Silyl-modified Bellus-Claisen rearrangement.
PMID 17325811
Abstract
A silyl-modified variant of the Bellus-Claisen rearrangement is described; the generality of this rearrangement has been demonstrated with a range of allylic amines and ketenes.