EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Lack of effect of aciclovir on metabolism of theophylline and expression of hepatic cytochrome P450 1A2 in rats.
PMID 17329857
Abstract
There is an interesting clinical report indicating that aciclovir, which is mainly excreted into urine, decreases the systemic clearance of theophylline by inhibiting cytochrome P450 (CYP) 1A2-mediated metabolism. In this study, we investigated the effect of aciclovir on the metabolism of theophylline, and on the activity and expression of hepatic CYP1A2 in rats. Theophylline (10 mg/kg) was injected intravenously into rats treated with two different dosages of aciclovir. When theophylline was simultaneously administered with aciclovir (50 mg/kg), the systemic clearance of theophylline and metabolic clearance of its major metabolites, 1-methyluric acid and 1,3-dimethyluric acid, were unchanged. In place of theophylline, when 1-methyl-3-propylxanthine (2.5 mg/kg), which is almost metabolized by CYP1A2 in rats, was coadministered intravenously with aciclovir (50 mg/kg), the pharmacokinetics of 1-methyl-3-propylxanthine was also unchanged. When theophylline was administered to rats pretreated with repeated intraperitoneal injections of aciclovir (25 mg/kg twice daily for 3 d), no significant differences in the systemic clearance of theophylline and its metabolic clearance to 1-methyluric acid and 1,3-dimethyluric acid were observed between the control and aciclovir-treated rats. This dosage of aciclovir did not change the activity of 7-ethoxyresorufin O-dealkylation, which is represented as CYP1A2 activity. In Western blot analysis, no significant change in the protein levels of hepatic CYP1A2 was observed between the control and aciclovir-treated rats. The present study suggests that aciclovir has no effect on the pharmacokinetics and metabolism of theophylline and on the activity and expression of hepatic CYP1A2 in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6885
1-Methyluric acid
C6H6N4O3