EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Four novel metabolites from microbial transformation of curcumol by Cunninghamella blakesleana.
PMID 17329889
Abstract
Further study on the microbial transformation of curcumol (1) by Cunninghamella blakesleana (AS3.970) led to the isolation of four novel metabolites. Their structures were elucidated as 3beta-hydroxy curcumol (2), 12-hydroxy curcumol (3), 1alpha-hydroxy-10beta,14-epoxy curcumol (4) and (2S,4S,5S,7S)-10-hydroxymethyl-7-isopropyl-2-methoxy-4-methyl-1-oxaspiro[4,6]undec-10-en-8-one (5) on the basis of spectral methods. All of them were characterized as new compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36236
( −)-Curcumol, analytical standard
C15H24O2