EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
Gene expression analyses of the liver in rats treated with oxfendazole.
PMID 17340121
Abstract
The effect of oxfendazole (OX), a benzimidazole anthelmintic, on hepatic gene expression was investigated in the liver of rats as a preliminary study to elucidate the possible mechanism of its non-genotoxic hepatocarcinogenesis. The liver from a male F344/N rat given a diet containing 500 ppm of OX for 3 weeks was examined by global gene expression analysis in comparison with an untreated rat. Microarray analysis revealed that phase I and phase II detoxifying enzymes were up-regulated in an OX-treated rat. In addition to these genes, the expressions of several upregulated genes related to xenobiotic metabolism and oxidative stress [e.g. Cyp1a1; NAD(P)H dehydrogenase, quinone 1 (Nqo1); glutathione peroxidase 2 (Gpx2); glutathione S-transferase Yc2 subunit (Yc2)], were confirmed by real-time reverse transcription polymerase chain reaction (RT-PCR). Furthermore, rats were administered 500 or 1,000 ppm of OX for 9 weeks, and the effect of OX on oxidative stress responses was evaluated by real-time RT-PCR along with conventional toxicological assays, including lipid peroxidation (thiobarbituric acid-reactive substance; TBARS). A longer treatment period and/or a higher dose of OX tended to increase the gene expressions of not only phase I (Cyp1a1 and Cyp1a2) but also phase II (Nqo1, Gpx2, Yc2, and Akr7a3) drug metabolizing enzymes. Toxicological parameters, such as TBARS, serum aspartate aminotransferase (AST), and serum alkaline phosphatase (ALP), showed slight but significant increases after treatment with OX for 9 weeks. These results indicate that OX elicits adaptive responses against oxidative stress in the liver and suggest that the imbalance in redox status might be one of the factors triggering the initial step of OX-induced non-genotoxic carcinogenesis in the liver of rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34176
Oxfendazole, VETRANAL, analytical standard
C15H13N3O3S