EMAIL THIS PAGE TO A FRIEND
Human Molecular Genetics
Leucine-rich repeat kinase 2 associates with lipid rafts.
PMID 17341485
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB9682
Anti-Dardarin Antibody, serum, clone, Chemicon®