EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis of 6- and 9-alkylaminomethyl furoflavones as gastroprotective agents.
PMID 17346862
Abstract
The synthesis of 9- and 6-alkylaminomethyl furoflavones 5a, b, 9a-c, 13a, b, 15a-g and 18 from the naturally occurring chromones visnagin and khellin. Gastroprotective potency of these compounds in the ethanol damage model was determined. The results indicate that, through appropriate substitution, furoflavones can be obtained that are gastroprotective.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60730
Khellin, for microscopy
C14H12O5