EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
High performance liquid chromatographic determination of 3-methylflavone-8-carboxylic acid, the main active metabolite of flavoxate hydrochloride in human urine.
PMID 17349765
Abstract
High performance liquid chromatographic (HPLC) method was presented for the determination of 3-methylflavone-8-carboxylic acid as the main active metabolite of flavoxate hydrochloride (FX) in human urine. The proposed method was based on using CN column with mobile phase consisting of acetonitrile-12 mM ammonium acetate (40:60, v/v) and adjusted to apparent pH 4.0 with flow rate of 1.5 ml min(-1). Quantitation was achieved with UV detection at 220 nm. The proposed method was utilized to the determination of dissolution rate for tablets containing flavoxate hydrochloride. The urinary excretion pattern has been calculated using the proposed method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8304
Flavoxate hydrochloride, ≥98% (HPLC), solid
C24H25NO4 · HCl