EMAIL THIS PAGE TO A FRIEND
Cancer research
Picropodophyllotoxin or podophyllotoxin does not induce cell death via insulin-like growth factor-I receptor.
PMID 17363615
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9576
Picropodophyllotoxin, ≥96% (HPLC), powder
C22H22O8