EMAIL THIS PAGE TO A FRIEND
International journal of toxicology
Effect of clebopride, antidopaminergic gastrointestinal prokinetics, on cardiac repolarization.
PMID 17365143
Abstract
The inhibition of the potassium current I(Kr) and QT prolongation has been known to be associated with drug-induced torsades de pointes arrhythmias (TdP) and sudden cardiac death. In this study, the authors investigated the cardiac electrophysiological effects of clebopride, a class of antidopaminergic gastrointestinal prokinetic, that has been reported to prolong the QT interval by using the conventional microelectrode recording techniques in isolated rabbit Purkinje fiber and whole-cell patch clamp techniques in human ether-à-go-go-related gene (hERG)-stably transfected Chinese hamster ovarian (CHO) cells. Clebopride at 10 microM significantly decreased the Vmax of phase 0 depolarization (p < .05) and significantly prolonged the action potential duration at 90% repolarization (APD90) (p < .01), whereas the action potential duration at 50% repolarization (APD50) was not prolonged. For hERG potassium channel currents, the IC50 value was 0.62 +/- 0.30 microM. Clebopride was found to have no effect on sodium channel currents. When these results were compared with Cmax (1.02 nM) of clinical dosage (1 mg, [p.o.]), it can be suggested that clebopride is safe at the clinical dosage of 1 mg from the electrophysiological aspect. These findings indicate that clebopride, an antidopaminergic gastrointestinal prokinetic drug, may provide a sufficient "safety factor" in terms of the electrophysiological threshold concentration. But, in a supratherapeutic concentration that might possibly be encountered during overdose or impaired metabolism, clebopride may have torsadogenic potency.