Journal of receptor and signal transduction research
Dominance of the alpha1B-adrenergic receptor and its subcellular localization in human and TRAMP prostate cancer cell lines.
PMID 17365508
The function and distribution of alpha1-adrenergic receptor (AR) subtypes in prostate cancer cells is well characterized. Previous studies have used RNA localization or low-avidity antibodies in tissue or cell lines to determine the alpha1-AR subtype and suggested that the alpha1A-AR is dominant. Two androgen-insensitive, human metastatic cancer cell lines DU145 and PC3 were used as well as the mouse TRAMP C1-C3 primary and clonal cell lines. The density of alpha1-ARs was determined by saturation binding and the distribution of the different alpha1-AR subtypes was examined by competition-binding experiments. In contrast to previous studies, the major alpha1-AR subtype in DU145, PC3 and all of the TRAMP cell lines is the alpha1B-AR. DU145 cells contained 100% of the alpha1B-AR subtype, whereas PC3 cells were composed of 21% alpha1 A-AR and 79% alpha1B-AR. TRAMP cell lines contained between 66% and 79% of the alpha1B-AR with minor fractions of the other two subtypes. Faster doubling time in the TRAMP cell lines correlated with decreasing alpha 1B-AR and increasing alpha1 A- and alpha1D-AR densities. Transfection with EGFP-tagged alpha1B-ARs revealed that localization was mainly intracellular, but the majority of the receptors translocated to the cell surface after extended preincubation (18 hr) with either agonist or antagonist. Localization was confirmed by ligand-binding studies and inositol phosphate assays where prolonged preincubation with either agonist and/or antagonist increased the density and function of alpha 1-ARs, suggesting that the native receptors were mostly intracellular and nonfunctional. Our studies indicate that alpha1B-ARs are the major alpha1-AR subtype expressed in DU145, PC3, and all TRAMP cell lines, but most of the receptor is localized in intracellular compartments in a nonfunctional state, which can be rescued upon prolonged incubation with any ligand.
Related Materials
Product #
Molecular Formula
Add to Cart
5-Methylurapidil, solid