EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Isolation of high-purity casticin from Artemisia annua L. by high-speed counter-current chromatography.
PMID 17374377
Abstract
Following an initial clean-up step on silica, high-speed counter-current chromatography (HSCCC) was used to purify a flavone, casticin (5,3'-dihydroxy-3,6,7, 4'-tetramethoxyflavone), from an extract of the dried leaves of Artemisia annua L. The two-phase solvent system used was composed of n-hexane-ethyl acetate-methanol-water at an optimized volume ratio of 7:10:7:10 (v/v). HSCCC separation of 226.4 mg of crude sample (containing casticin at 16.5% purity after silica gel clean-up) yielded 36.3 mg of casticin with a purity of over 99% and 96.2% recovery. Identification of the target compound was performed by (1)H NMR, (13)C NMR, two-dimensional NMR, electrospray ionization MS, IR and UV.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0189 Casticin, from Vitex trifolia, ≥98% (HPLC)
C19H18O8
16382
Casticin, analytical standard
C19H18O8