EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and crystallographic analysis of benzophenone derivatives--the potential anti-inflammatory agents.
PMID 17376692
Abstract
Fries rearrangement of substituted phenyl benzoates 1a-j to substituted hydroxy benzophenones 2a-j was achieved in excellent yield. Further benzoylation of 2a-j to benzoyloxy benzophenones 4a-n, a benzophenone analogue was achieved in good yield. All the newly synthesized compounds were evaluated for their anti-inflammatory activity and were compared with standard drugs. Out of the compounds studied, the compounds 4c, 4e, 4g, 4h and 4k with chloro and methyl substituents at para position showed more potent activity than the standard drugs at all doses tested.