EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
A theoretical investigation on the geometry and vibrational spectra of 10,10,2,6,5-pentamethyl-1-hydroxychroman: a model of alpha-tocopherol.
PMID 17376735
Abstract
In the present study, density functional theory calculations with the combined Becke's three-parameter exchange functional in combination with the Lee, Yang, and Parr correlation functional (B3LYP) exchange-correlation energy functions were performed by using the 6-311G** basis set to study the structure and vibrational spectra of 10,10,2,6,5-pentamethyl-1-hydroxychroman (a model of alpha-tocopherol). The fully optimized geometry of the molecule was found to be very consistent with the X-ray crystal structure. The predicted vibrational frequencies made it possible to give a reliable assignment of the IR spectrum of the molecule according to the potential energy distributions (PEDs).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
430676
2,2,5,7,8-Pentamethyl-6-chromanol, 97%
C14H20O2