EMAIL THIS PAGE TO A FRIEND
Nature
Organic chemistry: synthesis undressed.
PMID 17377569
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T89605
Tropinone, 99%
C8H13NO