EMAIL THIS PAGE TO A FRIEND
Nature
Biochemistry: molecular cannibalism.
PMID 17377573
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D147206
5,6-Dimethylbenzimidazole, ≥99%
C9H10N2