EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Alpha-phenyl-N-tert-butyl nitrone (PBN) derivatives: synthesis and protective action against microvascular damages induced by ischemia/reperfusion.
PMID 17379527
Abstract
Nitrones 4-7, structurally related to PBN (1), were prepared by reaction of the corresponding aromatic aldehydes with N-tert-butyl hydroxylamine. The protective effects of these nitrones against microvascular damages in ischemia/reperfusion in the 'hamster cheek pouch' assay were studied and 1, as well as 4a, 4b, and 7 (derived from piperonal, O-benzyl vanillin, and furfural, respectively), showed to be more active than shark cartilage or alpha-tocopherol. No correlation was found between the protective effect of these nitrones and their logP (partition coefficient) or their capacity to trap (*)OH and (*)CH(3) radicals.