EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
One pot synthesis of pyrimidine and bispyrimidine derivatives and their evaluation for anti-inflammatory and analgesic activities.
PMID 17383186
Abstract
A number of pyrimidine derivatives (1-10) have been synthesized by condensation of 4-isothiocyanato-4-methylpentan-2-one with furfurylamine, histamine, 1-(3-aminopropyl)imidazole, 1-(3-aminopropyl)-2-pyrrolidinone, 2-aminobenzonitrile and 3-isothiocyanatobutanal with 1-(3-aminopropyl)-2-pyrrolidinone and 2-hydrazinopyridine under different reaction conditions. Various bispyrimidine derivatives (11-15) were obtained by condensation of 4-isothiocyanato-4-methylpentan-2-one with 2,4,8,10-tetraoxaspiro[5,5]undecane3,9-dipropamine (11'), 1,4-bis(3-aminopropyl)piperazine (13'), 3,5-diamino 1,2,4-triazole (15') and 3-isothiocyanatobutanal with 2,4,8,10-tetraoxaspiro[5,5]undecane 3,9-dipropamine, 1,4-bis(3-aminopropyl)piperazine. All these compounds were characterized by correct FT-IR, (1)H NMR, MS and elemental analysis. These compounds were screened for anti-inflammatory and analgesic activities. Anti-inflammatory activity of 3 is comparable while analgesic activity was found to be better than that of standard drug.