EMAIL THIS PAGE TO A FRIEND
Biopolymers
Fmoc-2-mercaptobenzothiazole, for the introduction of the Fmoc moiety free of side-reactions.
PMID 17385207
Abstract
A double side-reaction, consisting in the formation of Fmoc-beta-Ala-OH and Fmoc-beta-Ala-AA-OH, during the preparation of Fmoc protected amino acids (Fmoc-AA-OH) with Fmoc-OSu is discussed. Furthermore, the new Fmoc-2-MBT reagent is proposed for avoiding these side-reactions as well as the formation of the Fmoc-dipeptides (Fmoc-AA-AA-OH) and even tripeptides, which is another important side-reaction when chloroformates such as Fmoc-Cl is used for the protection of the alpha-amino function of the amino acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3302
2-Mercaptobenzothiazole, 97%
C7H5NS2
63730
2-Mercaptobenzothiazole, technical, ≥90% (T)
C7H5NS2