EMAIL THIS PAGE TO A FRIEND
Chemical Society reviews
Functional ruthenium(II)- and iridium(III)-containing polymers for potential electro-optical applications.
PMID 17387410
Abstract
The need for novel materials with luminescent properties and advanced processing features requires reliable and reproducible synthetic routes for the design of suitable materials, such as e.g. polypyridyl ruthenium(II) and iridium(III)-containing polymers. The most popular ligand for those purposes is the 4,4'-functionalized bipyridine unit. Therefore, several synthetic strategies for the derivatization of the 4,4'-dimethyl-2,2'-bipyridine are highlighted, and in particular functionalities, which enable further covalent linkage to polymeric structures, are discussed in this critical review. Subsequently, the different synthetic strategies for the preparation of polymeric metal-complexes are described, either starting from small functionalized complexes (later covalently attached to the polymer), or from macroligands (subsequently coordinated to the metal ions). The designed materials reveal good processing properties using spin coating and inkjet printing, as well as beneficial electro-optical properties for potential thin functional film applications, such as light-emitting electrochemical cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209686
Iridium, powder, 99.9% trace metals basis
Ir
449229
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Ir
357324
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Ir
266841
Iridium, wire, diam. 0.15 mm, 99.9% trace metals basis
Ir
GF26117490
Iridium, wire, straight, 100mm, diameter 0.5mm, as drawn, 99.9%
Ir
GF35321172
Iridium, rod, 50mm, diameter 3.0mm, as drawn, 99.9%
Ir
GF96543209
Iridium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Ir
GF50717156
Iridium, wire, straight, 100mm, diameter 0.8mm, as drawn, 99.9%
Ir
GF98486982
Iridium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Ir
GF82950273
Iridium, wire reel, 0.2m, diameter 0.25mm, as drawn, 99.9%
Ir
GF17010986
Iridium, wire, straight, 100mm, diameter 1.0mm, as drawn, 99.9%
Ir
GF36309450
Iridium, wire reel, 0.1m, diameter 0.38mm, as drawn, 99.9%
Ir
GF31863607
Iridium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9%
Ir
336793
Iridium, black, powder, −200 mesh, ≥99% trace metals basis
Ir
GF07750795
Iridium, wire reel, 0.025m, diameter 2.0mm, as drawn, 99.9%
Ir
GF44052667
Iridium, wire reel, 0.05m, diameter 0.075mm, as drawn, 99.9%
Ir
GF97286428
Iridium, wire reel, 0.05m, diameter 0.1mm, as drawn, 99.9%
Ir
GF22425375
Iridium, wire reel, 0.05m, diameter 0.8mm, as drawn, 99.9%
Ir
GF68849544
Iridium, wire reel, 0.05m, diameter 2.0mm, as drawn, 99.9%
Ir
GF06583394
Iridium, wire reel, 0.1m, diameter 0.075mm, as drawn, 99.9%
Ir
GF65585021
Iridium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.5%
Ir
GF88210447
Iridium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Ir
GF26409247
Iridium, wire reel, 0.1m, diameter 0.15mm, as drawn, 99.9%
Ir
GF60803412
Iridium, wire reel, 0.1m, diameter 0.1mm, as drawn, 99.9%
Ir
GF99945534
Iridium, wire reel, 0.1m, diameter 0.2mm, as drawn, 99.9%
Ir
GF50407062
Iridium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Ir
GF80911403
Iridium, wire reel, 0.1m, diameter 0.8mm, as drawn, 99.9%
Ir
GF22923974
Iridium, wire reel, 0.2m, diameter 0.075mm, as drawn, 99.9%
Ir
GF30370924
Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.5%
Ir
GF26305130
Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.9%
Ir