EMAIL THIS PAGE TO A FRIEND
Annals of neurology
Mesenchymal stem cells effectively modulate pathogenic immune response in experimental autoimmune encephalomyelitis.
PMID 17387730
Abstract
To evaluate the ability of mesenchymal stem cells (MSCs), a subset of adult stem cells from bone marrow, to cure experimental autoimmune encephalomyelitis. The outcome of the injection of MSCs, in mice immunized with the peptide 139-151 of the proteolipid protein (PLP), was studied analyzing clinical and histological scores of treated mice. The fate of MSCs labeled with the green fluorescent protein was tracked in vivo by a photon emission imaging system and postmortem by immunofluorescence. The modulation of the immune response against PLP was studied through the analysis of in vivo T- and B-cell responses and by the adoptive transfer of MSC-treated encephalitogenic cells. MSC-treated mice showed a significantly milder disease and fewer relapses compared with control mice, with decreased number of inflammatory infiltrates, reduced demyelination, and axonal loss. In contrast, no evidence of green fluorescent protein-labeled neural cells was detected inside the brain parenchyma, thus not supporting the hypothesis of MSCs transdifferentiation. In vivo, PLP-specific T-cell response and antibody titers were significantly lower in MSC-treated mice. When adoptively transferred, encephalitogenic T cells activated against PLP(139-151) in the presence of MSCs induced a milder disease compared with that induced by untreated encephalitogenic T cells. These cells showed decreased production of interferon-gamma and tumor necrosis factor-alpha and did not proliferate on antigen recall, and thus were considered anergic. Overall, these findings suggest that the beneficial effect of MSCs in experimental autoimmune encephalomyelitis is mainly the result of an interference with the pathogenic autoimmune response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9184
Dexamethasone, meets USP testing specifications
C22H29FO5