EMAIL THIS PAGE TO A FRIEND
Organic letters
Straightforward stereoselective synthesis of spiro-epoxyoxindoles.
PMID 17394349
Abstract
[reaction: see text] The in situ preparation of a sulfonium ylide reagent achieved the highly diastereoselective epoxidation of isatins, so that a new and straightforward access to biologically significant spiro-epoxyoxindoles is provided. The first investigations of an asymmetric version are reported with enantiopure sulfides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183075
1-Methylisatin, 97%
C9H7NO2