EMAIL THIS PAGE TO A FRIEND
Organic letters
Asymmetric synthesis of primary amines via the spiroborate-catalyzed borane reduction of oxime ethers.
PMID 17397177
Abstract
[reaction: see text] The enantioselective borane reduction of O-benzyloxime ethers to primary amines was studied under catalytic conditions using the spiroborate esters 5-10 derived from nonracemic 1,2-amino alcohols and ethylene glycol. Effective catalytic conditions were achieved using only 10% of catalyst 5 derived from diphenylvalinol in dioxane at 0 degrees C resulting in complete conversion to the corresponding primary amine in up to 99% ee.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
443328
N-Isopropyl-N-methyl-tert-butylamine, 98%
C8H19N