EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Polypyrrole-polyvinyl sulphonate film based disposable nucleic acid biosensor.
PMID 17397646
Abstract
Double stranded calf thymus deoxyribonucleic acid entrapped polypyrrole-polyvinyl sulphonate (dsCT-DNA-PPy-PVS) films fabricated onto indium-tin-oxide (ITO) coated glass plates have been used to detect organophosphates such as chlorpyrifos and malathion. These disposable dsCT-DNA-PPy-PVS/ITO bioelectrodes have been characterized using cyclic voltammetry, Fourier-transform-infra-red (FTIR) spectroscopy and atomic force microscopy (AFM), respectively. These biosensing electrodes have a response time of 30s, are stable for about 5 months when stored in desiccated conditions at 25 degrees C and can be used to amperometrically detect chlorpyrifos (0.0016-0.025 ppm) and malathion (0.17-5.0), respectively. The additive effect of these pesticides on the amperometric response of the disposable dsCT-DNA-PPy-PVS/ITO bioelectrodes has also been investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
271969
Poly(vinyl sulfate) potassium salt, average Mw ~170,000