EMAIL THIS PAGE TO A FRIEND
Journal of microbiological methods
Novel insoluble dye-labeled substrates for screening inulin-degrading microorganisms.
PMID 17399828
Abstract
Inulin hydrogels were used to prepare insoluble colored substrates for screening of inulin-degrading microorganisms. Methycrylated inulin, synthesized with glycidyl methacrylate, was crossed-linked by free radical polymerization producing an insoluble hydrogel. The inulin hydrogels were colored with one of three covalent dyes (Reactive blue 4, Reactive orange 14, Reactive red 120). The efficacy of the colored inulin hydrogels was determined by comparing their performance to insoluble, colored amylose, xylan and HE-cellulose in screening assays. Novel substrates (alone or in combination with other insoluble colored substrates) were successfully used for screening inulin-degrading microorganisms from sludge samples. Selected strains were identified using 16S rDNA gene partial sequencing; all of them belong to the genus Bacillus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
244813
Reactive Blue 4, Dye content 35 %
C23H14Cl2N6O8S2
R0378
Reactive Red 120
C44H24Cl2N14Na6O20S6