EMAIL THIS PAGE TO A FRIEND
Anais da Academia Brasileira de Ciencias
Insecticidal activity of 2-tridecanone against the cowpea weevil Callosobruchus maculatus (Coleoptera: Bruchidae).
PMID 17401472
Abstract
The effect of 2-tridecanone vapor on the cowpea weevil (Callosobruchus maculatus) development was determined. Seeds of cowpea were infested with adults and exposed to different doses of 2-tridecanone isolated from Pilocarpus microphyllus Stapf ex Holm, a plant species native from northeastern Brazil. The pure monoterpene was evaluated both undiluted as well as in the dilutions 1:10, 1:100 and 1:1,000 (v/v). The following parameters of the cowpea weevil life cycle were analyzed in response to decreasing doses of 2-tridecanone: number of eggs laid, percentage of egg hatching on seeds, percentage of adult emergence, adult weight at emergence, mean developmental time and number of adults emerged. Vapor of 2-tridecanone caused a significant (P < 0.05) reduction in the number of eggs laid, in the percentage of eggs hatched and in the number of emerged adults in infested seeds. The fumigant insecticidal effect of 2-tridecanone was mainly due to its ovicidal activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W338818
2-Tridecanone, ≥96%, FG
C13H26O
172839
2-Tridecanone, 99%
C13H26O