EMAIL THIS PAGE TO A FRIEND
Haematologica
Mini-dose of thalidomide for treatment of primary myelofibrosis. Report of a case with complete reversal of bone marrow fibrosis and splenomegaly.
PMID 17405746
Abstract
We report a patient with very advanced myelofibrosis and huge splenomegaly who showed a complete hematological response to low dose thalidomide with reversal of splenomegaly and bone narrow fibrosis after 30 months of the treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7751
Fluoxymesterone, solid (photosensitive)
C20H29FO3