EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Plasma total coenzyme Q9 (CoQ9) in the New Zealand population: reference interval and biological variation.
PMID 17405953
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27597
Coenzyme Q9, ≥96.0% (HPLC)
C54H82O4