EMAIL THIS PAGE TO A FRIEND
Nature protocols
Thermal and high pressure intramolecular Diels-Alder reaction of vinylsulfonamides.
PMID 17406571
Abstract
Vinylsulfonamides with a furan, carbocyclic, semicyclic or acyclic 1,3-diene moiety are synthetized via a domino elimination-amidation reaction of 2-chloroethanesulfonyl chloride. Intramolecular Diels-Alder reaction of these vinylsulfonamides with thermal (toluene, 110 degrees C) or high pressure (dichloromethane, 13 kbar) activation provides efficient access to a range of gamma- and delta-sultams following a 2-3 d long synthetic procedure. Enantiopure sultams are readily obtained from N-1-phenylethyl substituted vinylsulfonamides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
302325
2-Chloroethanesulfonyl chloride, 96%
C2H4Cl2O2S