EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Rhodium-catalyzed asymmetric synthesis of 3,3-disubstituted 1-indanones.
PMID 17407122
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
661023
Hydroxy(cyclooctadiene)rhodium(I) dimer, 95%
C16H26O2Rh2