EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Novel series of substituted biphenylmethyl urea derivatives as MCH-R1 antagonists for the treatment of obesity.
PMID 17407817
Abstract
We have designed and synthesized two novel series of MCH-R1 antagonists based on a substituted biphenylmethyl urea core. SAR was explored, suggesting that optimal binding with the receptor was achieved when the biphenylmethyl group and the linker were substituted on the same nitrogen of the urea moiety. Compound 1-(3'-cyano-4-biphenylmethyl)-3-(2-hydroxy-1,1-dimethylethyl)-1-{2-[1-(4-methylbenzyl)-4-piperidinyl]ethyl}urea 2t showed the best antagonist binding activity to the MCH-R1 with a 43 nM K(i).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
479152
4-Bromobenzylamine, 96%
C7H8BrN