EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Ionic-liquid-promoted palladium-catalyzed multicomponent cyclocarbonylation of o-iodoanilines and allenes to form methylene-2,3-dihydro-1H-quinolin-4-ones.
PMID 17408285
Abstract
The palladium-catalyzed cyclocarbonylation reaction of o-iodoanilines with allenes and CO in 1-butyl-3-methylimidazolium hexafluorophosphate afforded 3-methylene-2,3-dihydro-1H-quinolin-4-ones in moderate to excellent yields under a low pressure (5 atm) of CO. The ionic liquid, as the solvent and promoter, enhances the efficiency of the cyclocarbonylation reaction. The recyclability of the system of ionic liquid/catalyst/ligand was also demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7209
3-Iodoaniline, 98%
C6H6IN