EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Effects of benzo(e)pyrene and benzo(a)pyrene on P-glycoprotein-mediated transport in Caco-2 cell monolayer: a comparative approach.
PMID 17408918
Abstract
The previous studies from our laboratory reported that benzo(a)pyrene (Bap) influenced efflux transport of rhodamine 123 (Rho-123) by induction of P-glycoprotein (P-gp) in Caco-2 cells. The present study investigated whether induction of P-gp and the enhanced efflux transport of Rho-123 were caused by benzo(e)pyrene (Bep), which has a structure similar to Bap, but is not a carcinogenic compound. In Caco-2 monolayer exposed to 50 microM Bep for 72 h, the ratio of the apparent permeability coefficient (P(app)) of Rho-123 efflux increased significantly compared to that of the control monolayer. Similarly, a significant increase in expression of MDR1 mRNA and of P-gp at the protein level were detected by RT-PCR and by Western blot analysis, respectively, in Caco-2 cells exposed to Bep, compared to that of the control. Caco-2 cells exposed to Bep showed oxidative stress that was detected by fluorescence microscopy using aminophenyl fluorescein. However, the oxidative stress was weaker compared with that of Bap. The cellular GSH content was decreased to 80% or 59% of control cells, respectively, in Caco-2 cells exposed to either Bep or Bap. Our results further show that Bep or Bap-induced P-gp in Caco-2 cells might have been the result of oxidative stress rather than DNA damage.