EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Addition of neuA, the gene encoding N-acylneuraminate cytidylyl transferase, increases the discriminatory ability of the consensus sequence-based scheme for typing Legionella pneumophila serogroup 1 strains.
PMID 17409215
Abstract
The standard sequence-based method for the typing of Legionella pneumophila serogroup 1 strains was extended by using the gspA and neuA alleles. The use of neuA as a seventh allele for typing significantly increased the index of discrimination calculated for a panel of unrelated strains (from 0.932 to 0.963) and subdivided some known large common complexes (e.g., 1,4,3,1,1,1). This modification to the standard method is proposed as the method of choice in the epidemiological investigation of L. pneumophila infections.