EMAIL THIS PAGE TO A FRIEND
Cancer research
Nonsteroidal anti-inflammatory drugs induce colorectal cancer cell apoptosis by suppressing 14-3-3epsilon.
PMID 17409426
Abstract
To determine the role of 14-3-3 in colorectal cancer apoptosis induced by nonsteroidal anti-inflammatory drugs (NSAIDs), we evaluated the effects of sulindac on 14-3-3epsilon protein expression in colorectal cancer cells. Sulindac sulfide inhibited 14-3-3epsilon proteins in HT-29 and DLD-1 cells in a time- and concentration-dependent manner. Sulindac sulfone at 600 mumol/L inhibited 14-3-3epsilon protein expression in HT-29. Indomethacin and SC-236, a selective cyclooxygenase-2 (COX-2) inhibitor, exerted a similar effect as sulindac. Sulindac suppressed 14-3-3epsilon promoter activity. As 14-3-3epsilon promoter activation is mediated by peroxisome proliferator-activated receptor delta (PPARdelta), we determined the correlation between 14-3-3epsilon inhibition and PPARdelta suppression by NSAIDs. Sulindac sulfide inhibited PPARdelta protein expression and PPARdelta transcriptional activity. Overexpression of PPARdelta by adenoviral transfer rescued 14-3-3epsilon proteins from elimination by sulindac or indomethacin. NSAID-induced 14-3-3epsilon suppression was associated with reduced cytosolic Bad with elevation of mitochondrial Bad and increase in apoptosis which was rescued by Ad-PPARdelta transduction. Stable expression of 14-3-3epsilon in HT-29 significantly protected cells from apoptosis. Our findings shed light on a novel mechanism by which NSAIDs induce colorectal cancer apoptosis via the PPARdelta/14-3-3epsilon transcriptional pathway. These results suggest that 14-3-3epsilon is a target for the prevention and therapy of colorectal cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S1438 Sulindac sulfone, ≥94% (HPLC), (solid or powder)
C20H17FO4S