EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Regioselective and enzymatic production of gamma-resorcylic acid from resorcinol using recombinant Escherichia coli cells expressing a novel decarboxylase gene.
PMID 17415666
Abstract
A recombinant Escherichia coli, expressing the rdc gene, which encodes a gamma-resorcylic acid decarboxylase (Rdc) reversibly catalyzing regioselective carboxylation of resorcinol derived from Rhizobium radiobacter WU-0108, converted 20 mM resorcinol to gamma-resorcylic acid with a 44% (mol/mol) yield at 30 degrees C for 7 h. The recombinant E. coli cells were recyclable at least five times for use as biocatalysts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D109606
2,6-Dihydroxybenzoic acid, 98%
C7H6O4