EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis, and evaluation of tetrahydroquinoline and pyrrolidine sulfonamide carbamates as gamma-secretase inhibitors.
PMID 17418571
Abstract
Gamma-secretase is a key enzyme involved in the production of beta-amyloid peptides which are believed to play a critical role in the onset and progression of Alzheimer's disease (AD). As such, inhibition of gamma-secretase has been an attractive approach to AD therapy. In this paper, the design, synthesis, and evaluation of tetrahydroquinoline and pyrrolidine sulfonamide carbamates as gamma-secretase inhibitors are described.