EMAIL THIS PAGE TO A FRIEND
Cell
HIF-1 regulates cytochrome oxidase subunits to optimize efficiency of respiration in hypoxic cells.
PMID 17418790
Abstract
O(2) is the ultimate electron acceptor for mitochondrial respiration, a process catalyzed by cytochrome c oxidase (COX). In yeast, COX subunit composition is regulated by COX5a and COX5b gene transcription in response to high and low O(2), respectively. Here we demonstrate that in mammalian cells, expression of the COX4-1 and COX4-2 isoforms is O(2) regulated. Under conditions of reduced O(2) availability, hypoxia-inducible factor 1 (HIF-1) reciprocally regulates COX4 subunit expression by activating transcription of the genes encoding COX4-2 and LON, a mitochondrial protease that is required for COX4-1 degradation. The effects of manipulating COX4 subunit expression on COX activity, ATP production, O(2) consumption, and reactive oxygen species generation indicate that the COX4 subunit switch is a homeostatic response that optimizes the efficiency of respiration at different O(2) concentrations. Thus, mammalian cells respond to hypoxia by altering COX subunit composition, as previously observed in yeast, but by a completely different molecular mechanism.