EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis and spectroscopic properties of lanthanide nitrate complexes with a new amide-based quinoxaline-2,3-dione ligand.
PMID 17419093
Abstract
Solid complexes of lanthanide nitrate with 1,4-di(N,N-di-n-butyl-acetamido)-quinoxaline-2,3-dione (L), [Ln(N03)3L.H2O] (Ln=La, Nd, Eu, Gd, Tb, Er), have been prepared and characterized by elemental analysis, IR, UV-vis spectra and conductivity measurements. The fluorescence property of the europium complex in solid state and in MeCN, acetone, AcOEt and THF was studied. Under the excitation, the europium complex exhibited characteristic emissions of europium. The result indicates that the triplet state energy level of the ligand matches better to the resonance level of Eu(III) than Tb(III) ion.
Related Materials