EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Inhibitory activities of three classes of acyclic nucleoside phosphonates against murine polyomavirus and primate simian virus 40 strains.
PMID 17420214
Abstract
Murine polyomavirus and simian virus 40 were used to evaluate the potencies of the compounds of three classes of acyclic nucleoside phosphonates: (i) the original HPMP (3-hydroxy-2-phosphonomethoxypropyl) and PME (2-phosphonomethoxyethyl) derivatives, (ii) the 6-[2-(phosphonomethoxy)alkoxy]-2,4-diaminopyrimidine (DAPy) derivatives, and (iii) a new class of HPMP derivatives containing a 5-azacytosine moiety. The last class showed the highest activities and selectivities against both polyomaviruses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
468231
2,4-Diaminopyrimidine, 98%
C4H6N4