EMAIL THIS PAGE TO A FRIEND
Journal of veterinary internal medicine
Neurotoxicosis in 4 cats receiving ronidazole.
PMID 17427396
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R7635
Ronidazole, analytical standard
C6H8N4O4