EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Organic-inorganic hybrids based on pyrene functionalized octavinylsilsesquioxane cores for application in OLEDs.
PMID 17428057
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
391573
1-Bromopyrene, 96%
C16H9Br